Learn More
Medicus Health Mega Test Tube Racks, 10-13Mm, Holds 432 Test Tubes, White
SDP
Catalog No. NC1938646
Encompass
∙ Mega Test Tube Racks hold 432 total 10-13mm test tubes for high volume testing and storage ∙Now available in Blue or White so you can separate tests and storage by color ∙Mega Racks Racks are perfect for organizing and storing specimen tubes in any size Laboratory∙Mega Racks hold 432 total 10-13mm test tubes for high volume testing and storage∙Max size is 5-7 mL Test Tubes ∙Rack has identifying alpha-numeric for quick location and retrieval of tubes∙Tubes are spaced far enough apart to make finding repeats, put always and send outs a snap∙Made of autoclavable polypropylene∙Overall Dimensions: 26.5"L x 10.5"W x 3.5"H
Catalog No. NC1938646 Supplier Medicus Health Supplier No. 5291M7WHITE
∙ Mega Test Tube Racks hold 432 total 10-13mm test tubes for high volume testing and storage ∙Now available in Blue or White so you can separate tests and storage by color ∙Mega Racks Racks are perfect for organizing and storing specimen tubes in any size Laboratory∙Mega Racks hold 432 total 10-13mm test tubes for high volume testing and storage∙Max size is 5-7 mL Test Tubes ∙Rack has identifying alpha-numeric for quick location and retrieval of tubes∙Tubes are spaced far enough apart to make finding repeats, put always and send outs a snap∙Made of autoclavable polypropylene∙Overall Dimensions: 26.5"L x 10.5"W x 3.5"H