Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ MegaClean™ Heavy-Duty Cleaner
BIOPHARMASDP

Catalog No. 19306970 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-306-970 5 gal.
19-035-380E 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-306-970 Supplier Micronova™ Supplier No. MC15G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.

The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.

  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations

Specifications

Product Type MegaClean Cleaner
Color-coded No
Handle Type None
Sterility Non-sterile
Description Heavy Duty Detergent, Degreaser
For Use With (Application) Ideal spot cleaning solution and easily removes grease, adhesives and photo-resist stains from most surfaces, floors, high traffic areas, parts cleaning, adhesive removal
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.