Learn More
STEMCELL Technologies MegaCult™-C Collagen and Medium with Lipids, Size: 1 Kit
SDP
Catalog No. 501974337
Encompass
MegaCult™-C Collagen and Medium with Lipids kit includes medium and collagen solution for colony-forming unit (CFU) assays of human or mouse megakaryocyte progenitor cells (CFU-Mk). The addition of cytokines is required.
Catalog No. 50-197-4337 Supplier STEMCELL Technologies Supplier No. 04974
MegaCult™-C Collagen and Medium with Lipids kit includes medium and collagen solution for colony-forming unit (CFU) assays of human or mouse megakaryocyte progenitor cells (CFU-Mk). The addition of cytokines is required.