Learn More
STEMCELL Technologies MegaCult™-C Collagen and Medium with Cytokines, Size: 1 Kit
SDP
Catalog No. 501974333
Encompass
MegaCult™-C Collagen and Medium with Cytokines kit includes includes medium and collagen solution necessary for performing colony-forming unit (CFU) assays of human megakaryocyte progenitor cells (CFU-Mk). • MegaCult™-C Medium with Cytokines (/products/megacult-c-medium-with-cytokines.html) is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples. It is suitable for use with CD34 + enriched cells, mononuclear cells, and cells isolated by other purification methods. The medium contains recombinant human (rh) IL-3, IL-6, and thrombopoietin.• Collagen Solution (/products/collagen-solution.html) for preparation of collagen gels and for coating cell culture surfaces.Individual kit components may also be purchased separately, for your convenience.For more information on protocols for human CFU-Mk assays using MegaCult™-C, please explore the Technical Manual (#section-protocols-and-documentation).
Catalog No. 50-197-4333 Supplier STEMCELL Technologies Supplier No. 04961
MegaCult™-C Collagen and Medium with Cytokines kit includes includes medium and collagen solution necessary for performing colony-forming unit (CFU) assays of human megakaryocyte progenitor cells (CFU-Mk). • MegaCult™-C Medium with Cytokines (/products/megacult-c-medium-with-cytokines.html) is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples. It is suitable for use with CD34 + enriched cells, mononuclear cells, and cells isolated by other purification methods. The medium contains recombinant human (rh) IL-3, IL-6, and thrombopoietin.• Collagen Solution (/products/collagen-solution.html) for preparation of collagen gels and for coating cell culture surfaces.Individual kit components may also be purchased separately, for your convenience.For more information on protocols for human CFU-Mk assays using MegaCult™-C, please explore the Technical Manual (#section-protocols-and-documentation).