Learn More
STEMCELL Technologies MegaCult™-C Complete Kit with Cytokines, Size: 1 Kit
SDP
Catalog No. 501974336
Encompass
MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining.MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples. It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods.
Catalog No. 50-197-4336 Supplier STEMCELL Technologies Supplier No. 04971
Add to Cart
Add to Cart
Request a Quote
MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining.MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples. It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods.