Learn More
STEMCELL Technologies MegaCult™-C Complete Kit Without Cytokines, Size: 1 Kit
SDP
Catalog No. 501974335
Encompass
MegaCult™-C Complete Kit Without Cytokines includes all reagents (except cytokines) required for colony-forming unit (CFU) assays of megakaryocyte progenitor cells (CFU-Mk) in a collagen-based culture medium, including the detection of human CFU-Mk colonies by immunocytochemical staining. The addition of appropriate cytokines, as chosen by the user, is required.
Catalog No. 50-197-4335 Supplier STEMCELL Technologies Supplier No. 04970
Add to Cart
Add to Cart
Request a Quote
MegaCult™-C Complete Kit Without Cytokines includes all reagents (except cytokines) required for colony-forming unit (CFU) assays of megakaryocyte progenitor cells (CFU-Mk) in a collagen-based culture medium, including the detection of human CFU-Mk colonies by immunocytochemical staining. The addition of appropriate cytokines, as chosen by the user, is required.