Learn More
STEMCELL Technologies MegaCult™-C Medium with Lipids, Size: 50 mL
SDP
Catalog No. 501974329
Encompass
MegaCult™-C Medium with Lipids is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples after addition of appropriate cytokines. It is recommended for use with CD34+-enriched cells, mononuclear cells, and cells isolated by other purification methods. It can also be used for CFU-MK assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow after addition of appropriate cytokines.
Catalog No. 50-197-4329 Supplier STEMCELL Technologies Supplier No. 04850
MegaCult™-C Medium with Lipids is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples after addition of appropriate cytokines. It is recommended for use with CD34+-enriched cells, mononuclear cells, and cells isolated by other purification methods. It can also be used for CFU-MK assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow after addition of appropriate cytokines.