Learn More
STEMCELL Technologies MegaCult™-C Staining Kit for CFU-Mk, Size: 1 Kit
SDP
Catalog No. 501974334
Encompass
Stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium using the reagents included in MegaCult™-C Staining Kit for CFU-Mk. • MegaCult™-C Primary Antibody• MegaCult™-C Control Antibody• MegaCult™-C Human Serum• MegaCult™-C Alkaline Phosphatase Substrate Tablets• MegaCult™-C Biotin-Conjugated Goat Anti-Mouse IgG• MegaCult™-C Evans Blue Stain• MegaCult™-C 10% BSAMegakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system. The kit is also suitable for the detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk).For your convenience, the kit components are also available for purchase individually, upon request.
Catalog No. 50-197-4334 Supplier STEMCELL Technologies Supplier No. 04962
Stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium using the reagents included in MegaCult™-C Staining Kit for CFU-Mk. • MegaCult™-C Primary Antibody• MegaCult™-C Control Antibody• MegaCult™-C Human Serum• MegaCult™-C Alkaline Phosphatase Substrate Tablets• MegaCult™-C Biotin-Conjugated Goat Anti-Mouse IgG• MegaCult™-C Evans Blue Stain• MegaCult™-C 10% BSAMegakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system. The kit is also suitable for the detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk).For your convenience, the kit components are also available for purchase individually, upon request.