Learn More
Bioss Megalin Polyclonal Antibody, PE Conjugated, 100ul
SDP
Catalog No. NC3711156
Encompass_Preferred
Acts together with cubilin to mediate HDL endocytosis (By similarity). May participate in regulation of parathyroid-hormone and para-thyroid-hormone-related protein release.
Catalog No. NC3711156 Supplier Bioss Supplier No. BS3909RPE
Acts together with cubilin to mediate HDL endocytosis (By similarity). May participate in regulation of parathyroid-hormone and para-thyroid-hormone-related protein release.