Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diagenode Inc MEGARUPTOR 3 SHEARING KIT 16SA

Catalog No. NC2030992
Encompass

  • High-quality DNA fragmentation
  • Compatible with Megaruptor 3 system
  • Sample capacity of 16 samples
  • DNA fragment size range from 5 kb to over 100 kb
  • Utilizes hydrodynamic shearing method
  • Contains Hydropore-Syringe assemblies and Hydro Tubes
  • Components are sterile for reliable performance
  • Designed for single-use consumables
  • Can be stored at room temperature
  • Application focus on DNA fragmentation, NGS, and long-read sequencing
  • Intended for research use only
  • High reproducibility with < 15% CV DNA shearing variability under specific conditions

Catalog No. NC2030992 Supplier Diagenode Inc Supplier No. E07010003
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • High-quality DNA fragmentation
  • Compatible with Megaruptor 3 system
  • Sample capacity of 16 samples
  • DNA fragment size range from 5 kb to over 100 kb
  • Utilizes hydrodynamic shearing method
  • Contains Hydropore-Syringe assemblies and Hydro Tubes
  • Components are sterile for reliable performance
  • Designed for single-use consumables
  • Can be stored at room temperature
  • Application focus on DNA fragmentation, NGS, and long-read sequencing
  • Intended for research use only
  • High reproducibility with < 15% CV DNA shearing variability under specific conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.