Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MegaScript™ T7 Transcription Kit Plus

Catalog No. A5762225
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1000 Reactions
25 Reactions
50 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A5762225 25 Reactions
A57622 50 Reactions
A57623 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5762225 Supplier Invitrogen™ Supplier No. A5762225
Add to Cart
Add to Cart

The MegaScript T7 Transcription Kit Plus contains all the reagents and buffers necessary for in vitro synthesis of large amounts of RNA, is suitable for full- or partially-modified nucleotide substitution to produce modified RNA, and enables the use of different cap analogs.

The MegaScript T7 Transcription Kit Plus contains all the reagents and buffers necessary for in vitro synthesis of large amounts of RNA. Each 20-μL reaction yields an average of 100 μg of RNA, while the scaled-up protocol yields up to 1 mg of RNA. This kit is suitable for full- or partially-modified-nucleotide substitution to produce modified RNA. It is also suitable for co-transcriptional capping with cap analogs to make capped RNA that can be translated into protein. Easily scale from RUO to GMP with the incorporation of TheraPure reagents, eliminating reoptimization during scale up.

Features of the MegaScript T7 Transcription Kit Plus include:

  • Optimized to achieve high mRNA yields (>5 mg/mL)
  • Flexible—allows use of modified nucleotides and/or cap analogs such as ARCA or CleanCap
  • Includes GFP control template
  • Made up of high quality reagents (TheraPure); eliminates the need to qualify new reagents during scale-up
  • Available in two sizes: 50 and 1000 reactions

The MegaScript T7 Transcription Kit Plus requires a linearized DNA template with a T7 RNA polymerase promoter to create >5 mg/mL of RNA. MEGAscript kits are intended for the synthesis of large amounts of unlabeled or low-specific-activity RNA for a variety of uses, including transfections, preclinical mRNA therapeutic mRNA studies, microinjections, in vitro translation, and RNA structure and function analysis. Note that this kit does not create capped RNA, which is necessary for translational studies. MEGAscript kits are not recommended for synthesis of high-specific-activity probes.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type HEK293, 293, HeLa, CHO-K1, CHO, Jurkat, Primary Human T Cells, Primary Cells, Stem Cells, iPSC, NIH-3T3, HepG2, A549, HUVEC
Final Product Type Synthetic mRNA
Format Animal Origin Free Kit
Isolation Technology Silica Spin Column
Purification Target mRNA
Sample Type mRNA, Total RNA
No. of Reactions 25 Reactions
Product Line MEGAscript
Product Type T7 In Vitro Transcription Kit
Quantity 25 Reactions
Promoter T7
Shipping Condition Dry Ice
Yield >100 μg
Starting Material PCR Products, Linearized DNA Plasmid
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.