Learn More
Medchemexpress LLC HY-B1834 10mg Medchemexpress, Megestrol CAS:3562-63-8 Purity:>98%
SDP
Catalog No. 501970168
Encompass_Preferred
Medchemexpress, HY-B1834 10mg Megestrol CAS:3562-63-8 Megestrol is a synthetic progestin and used for the treatment of anorexia, cachexia, or an unexplained significant weight loss in patients with an acquired immunodeficiency syndrome diagnosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0168 Supplier Medchemexpress LLC Supplier No. HYB183410MG
Medchemexpress, HY-B1834 10mg Megestrol CAS:3562-63-8 Megestrol is a synthetic progestin and used for the treatment of anorexia, cachexia, or an unexplained significant weight loss in patients with an acquired immunodeficiency syndrome diagnosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.