Learn More
Selleck Chemical LLC Megestrol Acetate S1304-1g
SDP
Catalog No. 503108255
Encompass_Preferred
Megestrol acetate (BDH1298 SC10363) is a synthetic progestogen used to treat breast cancer and loss of appetite
Catalog No. 50-310-8255 Supplier Selleck Chemical LLC Supplier No. S13041G
Megestrol acetate (BDH1298 SC10363) is a synthetic progestogen used to treat breast cancer and loss of appetite