Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Megestrol acetate | 595-33-5 | 99.7% | 384.51 | 25 MG
SDP

Catalog No. 5000347901
Encompass_Preferred

Megestrol acetate is a synthetic and orally active progesteronal agent. It is effective as an appetite stimulant for wasting syndromes such as cachexia and has the potential for HIV study. It also decreases nuclear and cytosol androgen receptors in human BPH tissue and downregulates the autophagic catabolic pathway.

  • Synthetic and orally active progesteronal agent
  • Effective as an appetite stimulant for wasting syndromes like cachexia
  • Potential for HIV study
  • Decreases nuclear and cytosol androgen receptors in human BPH tissue
  • Downregulates the autophagic catabolic pathway
  • Appearance: solid
  • Color: white to off-white
  • Solubility in DMSO: 20 mg/mL, requires ultrasonic

Catalog No. 50-003-47901 Supplier Medchemexpress LLC Supplier No. 25MGHY13676R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Megestrol acetate is a synthetic and orally active progesteronal agent. It is effective as an appetite stimulant for wasting syndromes such as cachexia and has the potential for HIV study. It also decreases nuclear and cytosol androgen receptors in human BPH tissue and downregulates the autophagic catabolic pathway.

  • Synthetic and orally active progesteronal agent
  • Effective as an appetite stimulant for wasting syndromes like cachexia
  • Potential for HIV study
  • Decreases nuclear and cytosol androgen receptors in human BPH tissue
  • Downregulates the autophagic catabolic pathway
  • Appearance: solid
  • Color: white to off-white
  • Solubility in DMSO: 20 mg/mL, requires ultrasonic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.