Learn More
Medchemexpress LLC Megestrol acetate-d3 | 162462-72-8 | 387.53 | 500 UG
SDP
Catalog No. 5000347957
Encompass_Preferred
Megestrol acetate-d3 is a deuterium-labeled version of Megestrol acetate, a synthetic and orally active progesteronal agent. It is utilized as an appetite stimulant for wasting syndromes and decreases nuclear and cytosol androgen receptors. This compound holds potential for HIV studies and downregulates the autophagic catabolic pathway.
  • Effective as an appetite stimulant for wasting syndromes such as cachexia
  • Decreases nuclear and cytosol androgen receptors in human BPH tissue
  • Potential for HIV study and downregulates the autophagic catabolic pathway
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-47957 Supplier Medchemexpress LLC Supplier No. HY13676S500UG
Megestrol acetate-d3 is a deuterium-labeled version of Megestrol acetate, a synthetic and orally active progesteronal agent. It is utilized as an appetite stimulant for wasting syndromes and decreases nuclear and cytosol androgen receptors. This compound holds potential for HIV studies and downregulates the autophagic catabolic pathway.
  • Effective as an appetite stimulant for wasting syndromes such as cachexia
  • Decreases nuclear and cytosol androgen receptors in human BPH tissue
  • Potential for HIV study and downregulates the autophagic catabolic pathway
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs