Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Megestrol Acetate, MP Biomedicals™

Catalog No. ICN19023505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19023505 5 g
ICN19023501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19023505 Supplier MP Biomedicals, Inc Supplier No. 0219023505
Add to Cart
Add to Cart

Chemical Identifiers

CAS 595-33-5
Molecular Formula C24H32O4
Molecular Weight (g/mol) 384.516
InChI Key RQZAXGRLVPAYTJ-GQFGMJRRSA-N
Synonym megestrol acetate, niagestin, megace es, megestryl acetate, megace, megeron, ovaban, ovarid, pallace, magestin
PubChem CID 11683
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC1=CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C

Specifications

Quantity 5 g
Formula Weight 384.5
Physical Form Solid
Chemical Name or Material Megestrol Acetate
Recommended Storage Store at room temperature.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.