Learn More
Medchemexpress LLC Megestrol acetate (Standard) | 595-33-5 | 98.5% | 384.51 | 50 MG
SDP
Catalog No. 5000351709
Encompass_Preferred
Megestrol acetate (Standard) is the analytical standard of Megestrol acetate, intended for research and analytical applications. It is a synthetic and orally active progesteronal agent. This product is effective as an appetite stimulant for wasting syndromes like cachexia and decreases nuclear and cytosol androgen receptors in human BPH tissue. It also has potential for HIV study and downregulates the autophagic catabolic pathway.
  • Effective as an appetite stimulant for wasting syndromes
  • Decreases nuclear and cytosol androgen receptors in human BPH tissue
  • Potential for HIV study
  • Downregulates the autophagic catabolic pathway
  • Suitable for qualitative, quantitative, and methodological research in HPLC, GC, and MS
Catalog No. 50-003-51709 Supplier Medchemexpress LLC Supplier No. HY13676R50MG
Megestrol acetate (Standard) is the analytical standard of Megestrol acetate, intended for research and analytical applications. It is a synthetic and orally active progesteronal agent. This product is effective as an appetite stimulant for wasting syndromes like cachexia and decreases nuclear and cytosol androgen receptors in human BPH tissue. It also has potential for HIV study and downregulates the autophagic catabolic pathway.
  • Effective as an appetite stimulant for wasting syndromes
  • Decreases nuclear and cytosol androgen receptors in human BPH tissue
  • Potential for HIV study
  • Downregulates the autophagic catabolic pathway
  • Suitable for qualitative, quantitative, and methodological research in HPLC, GC, and MS