Learn More
Aviva Systems Biology Corp Megf6 Antibody - middle region (ARP45090_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45090_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Megf6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Megf6. Alternate names: Egfl3ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-183