Learn More
Cayman Chemical Meglutol-d3 10mg
SDP
Catalog No. 502991939
Encompass_Preferred
An internal standard for the quantification of meglutol by GC- or LC-MS
Catalog No. 50-299-1939 Supplier Cayman Chemical Supplier No. 3091410 MG
An internal standard for the quantification of meglutol by GC- or LC-MS