Learn More
Cayman Chemical Meglutol-d3 25mg
SDP
Catalog No. 502991386
Encompass_Preferred
An internal standard for the quantification of meglutol by GC- or LC-MS
Catalog No. 50-299-1386 Supplier Cayman Chemical Supplier No. 3091425 MG
Add to Cart
Add to Cart
An internal standard for the quantification of meglutol by GC- or LC-MS