Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meglutol-d3 (dicrotalic acid-d3) | 59060-36-5 | MFCD01074174 | 98.0% | 165.16 | C6H7D3O5 | 10 MG
SDP

Catalog No. 5000297399
Encompass_Preferred

Meglutol-d3 (dicrotalic acid-d3) is the deuterium-labeled analogue of meglutol, intended for use as a tracer or internal standard in quantitative analysis by NMR, GC-MS, or LC-MS. It is a white to off-white solid with high purity and is supplied in small-mass packages for analytical use.

  • Deuterium-labeled internal standard for quantitative analysis
  • Purity 98.0%
  • Molecular weight 165.16
  • Soluble in methanol (100 mg/mL; may require ultrasonic)
  • Stored as powder at -20°C (long term) or 4°C (short term)

Catalog No. 50-002-97399 Supplier Medchemexpress LLC Supplier No. HYB1189S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meglutol-d3 (dicrotalic acid-d3) is the deuterium-labeled analogue of meglutol, intended for use as a tracer or internal standard in quantitative analysis by NMR, GC-MS, or LC-MS. It is a white to off-white solid with high purity and is supplied in small-mass packages for analytical use.

  • Deuterium-labeled internal standard for quantitative analysis
  • Purity 98.0%
  • Molecular weight 165.16
  • Soluble in methanol (100 mg/mL; may require ultrasonic)
  • Stored as powder at -20°C (long term) or 4°C (short term)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.