Learn More
Lonza Walkersville MEGM Mammary Epithelial Cell Growth Medium
Catalog No. NC9523177
Encompass
Culture system containing MEBM Basal Medium (CC-3151) and MEGM SingleQuots Supplements (CC-4136) required for growth of Mammary Epithelial Cells.
Catalog No. NC9523177 Supplier Lonza Walkersville Supplier No. CC3150
Culture system containing MEBM Basal Medium (CC-3151) and MEGM SingleQuots Supplements (CC-4136) required for growth of Mammary Epithelial Cells.