Learn More
Lonza Walkersville MEGM Mammary Epithelial Cell Growth Medium SingleQuots Kit
Catalog No. NC9742592
Encompass
MEGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the MEGM Mammary Epithelial Cell Growth Medium BulletKit (Catalog Number CC-3150). The full MEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3150.Mammary Epithelial Cell Growth Medium BulletKit has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research. Content:1 x Orange Cap Vial with BPE 2.00 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL
Catalog No. NC9742592 Supplier Lonza Walkersville Supplier No. CC4136
MEGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the MEGM Mammary Epithelial Cell Growth Medium BulletKit (Catalog Number CC-3150). The full MEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3150.Mammary Epithelial Cell Growth Medium BulletKit has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research. Content:1 x Orange Cap Vial with BPE 2.00 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL