Learn More
Cole-Parmer AEMC 5050 Industrial Digital Megohmmeter, 5000 V
Supplier:  Cole-Parmer 2744310
Encompass
Measure insulation voltage levels up to 1000 or even to 5000 V. The programmable memory stores resistance values for specified time intervals. The backlit LCD features a dual display with a digital measurement and a bar graph to monitor changes or trends in readings. Automatic test inhibitor shuts down meter when live circuits exceed 25V — protecting the operator and equipment. The rugged case features a weatherproof design for heavy duty conditions.
Catalog No. 50-992-637