Learn More
Cayman Chemical MEG (sulfate), Purity: ≥98%, CAS: 3979-00-8, 50 mg
SDP
Catalog No. 502897011
Encompass_Preferred
MEG is a selective iNOS inhibitor and scavenger of peroxynitrite. The EC50 values for inhibition of iNOS (LPS-treated rat lung), eNOS (bovine endothelial), and nNOS (rat brain) are 11.5, 110, and 60 µM, respectively.{7417} MEG reacts with peroxynitrite with a second order rate constant of 1,900 M−1s−1 at 37°C.{4278} MEG has potent anti-inflammatory activity, which may be due to the combined effect of MEG acting as an inhibitor of iNOS and as a scavenger of peroxynitrite.{5922}
Catalog No. 50-289-7011 Supplier Cayman Chemical Supplier No. 8102050 MG
MEG is a selective iNOS inhibitor and scavenger of peroxynitrite. The EC50 values for inhibition of iNOS (LPS-treated rat lung), eNOS (bovine endothelial), and nNOS (rat brain) are 11.5, 110, and 60 µM, respectively.{7417} MEG reacts with peroxynitrite with a second order rate constant of 1,900 M−1s−1 at 37°C.{4278} MEG has potent anti-inflammatory activity, which may be due to the combined effect of MEG acting as an inhibitor of iNOS and as a scavenger of peroxynitrite.{5922}