Learn More
Cayman Chemical MEGXhydrochlorIde 10mg
SDP
Catalog No. 502941388
Encompass_Preferred
An active metabolite of lidocaine; formed via N-deethylation of lidocaine by hepatic CYP3A4; topical administration reduces thrombus formation in a hamster model of laser-induced microvascular injury at 2% v/v; plasma levels following lidocaine administration have been used to monitor declining liver function in patients with cirrhosis
Catalog No. 50-294-1388 Supplier Cayman Chemical Supplier No. 2801910 MG
An active metabolite of lidocaine; formed via N-deethylation of lidocaine by hepatic CYP3A4; topical administration reduces thrombus formation in a hamster model of laser-induced microvascular injury at 2% v/v; plasma levels following lidocaine administration have been used to monitor declining liver function in patients with cirrhosis