Learn More
MOLTOX Molecular Toxicology Inc mEI Agar, 60 mm Plate, 10-12 mL/plate, 10 plates/sleeve, Store in Refrigerator/Dark
SDP
Catalog No. 50948656
Encompass_Preferred
mEI Agar is a selective, chromogenic culture medium used for the detection and enumeration of enterococci in water via one-step membrane filtration technique. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-656 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S203
mEI Agar is a selective, chromogenic culture medium used for the detection and enumeration of enterococci in water via one-step membrane filtration technique. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.