Learn More
Meiji Techno America MEIJI EMZ-5TR-PLS-2/10 SCOPE
Catalog No. NC0885384
Encompass_Preferred
Trinocular Zoom Stereo microscope body with zoom range, W.D. 93mm 1,630.00 magnification from 7X to 90X (Using SWF10X and SWF20X eyepieces)
Catalog No. NC0885384 Supplier Meiji Techno America Supplier No. EMZ5TR
Trinocular Zoom Stereo microscope body with zoom range, W.D. 93mm 1,630.00 magnification from 7X to 90X (Using SWF10X and SWF20X eyepieces)