Learn More
Perkin Elmer US LLC MEINHARD Concentric Glass Nebulizer, Type K1
Catalog No. 502104213
Encompass_Preferred
The MEINHARD concentric glass nebulizer reflects exacting standards in design and manufacturing. Its advantages include simplicity of design and operation, reproducible and self-aligning aerosol, and close fabrication control. They provide excellent sensitivity and precision for aqueous solutions and samples with few dissolved solids (less than 1%). It includes Standard Nebulizer Solution Fitting, 0.5 mm PTFE capillary tubing (N0777414), PFA Gas Quick Connect (N8121036), and Nebulizer Clip for MEINHARD Nebulizers (N0777460).
Catalog No. 50-210-4213 Supplier Perkin Elmer US LLC Supplier No. N0777707
The MEINHARD concentric glass nebulizer reflects exacting standards in design and manufacturing. Its advantages include simplicity of design and operation, reproducible and self-aligning aerosol, and close fabrication control. They provide excellent sensitivity and precision for aqueous solutions and samples with few dissolved solids (less than 1%). It includes Standard Nebulizer Solution Fitting, 0.5 mm PTFE capillary tubing (N0777414), PFA Gas Quick Connect (N8121036), and Nebulizer Clip for MEINHARD Nebulizers (N0777460).