Learn More
Perkin Elmer US LLC MEINHARD Concentric Glass Nebulizer, Type K1 LDV
Catalog No. 502111771
Encompass_Preferred
The MEINHARD concentric nebulizers reflect exacting standards in design and manufacturing. They provide excellent sensitivity and precision for aqueous solutions and samples with few dissolved solids (less than 1%).
Catalog No. 50-211-1771 Supplier Perkin Elmer US LLC Supplier No. N0811287
The MEINHARD concentric nebulizers reflect exacting standards in design and manufacturing. They provide excellent sensitivity and precision for aqueous solutions and samples with few dissolved solids (less than 1%).