Learn More
Meinhard Elemental Scientific Meinhard HEN-120-A0.2 Nebulizer, High Efficiency, Borosilicate Glass, Operating Pressure 120 psi
Catalog No. NC0856674
Encompass
The Meinhard HEN-120-A0.2 is a high-efficiency glass nebulizer optimized for demanding analytical applications like ICP-MS. Its borosilicate glass construction and lapped flush nozzle design enhance its performance, allowing it to produce smaller aerosol droplets and operate effectively at low sample flow rates. With a nominal pressure of 120 psi and an aspiration rate of 200 µL/min, this nebulizer excels in sensitivity for analyzing small-volume samples in single-cell and particle applications.
Catalog No. NC0856674 Supplier Meinhard Elemental Scientific Supplier No. HEN120A0.2
The Meinhard HEN-120-A0.2 is a high-efficiency glass nebulizer optimized for demanding analytical applications like ICP-MS. Its borosilicate glass construction and lapped flush nozzle design enhance its performance, allowing it to produce smaller aerosol droplets and operate effectively at low sample flow rates. With a nominal pressure of 120 psi and an aspiration rate of 200 µL/min, this nebulizer excels in sensitivity for analyzing small-volume samples in single-cell and particle applications.