Learn More
Perkin Elmer US LLC MEINHARD Plus PFA Capillary Holder
Catalog No. 502111554
Encompass_Preferred
This Capillary Holder is also used with PerkinElmers High Throughput System (HTS). HTS affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. This holder is included in the Tubing Accessories Kit for HTS (N8150144).
Catalog No. 50-211-1554 Supplier Perkin Elmer US LLC Supplier No. N8152716
This Capillary Holder is also used with PerkinElmers High Throughput System (HTS). HTS affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. This holder is included in the Tubing Accessories Kit for HTS (N8150144).