Learn More
Aviva Systems Biology Corp Meis1 Antibody - N-terminal region (ARP38148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Meis1. It was validated on Western Blot. . Alternate names: C530044H18Rik, Evi8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-724