Learn More
Aviva Systems Biology Corp MEIS2 Antibody - N-terminal region (ARP34684_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34684_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MEIS2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MEIS2. Alternate names: MRG1, HsT18361ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55511