Learn More
Aviva Systems Biology Corp MEIS2 Antibody - N-terminal region (ARP36223_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MEIS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MEIS2. Alternate names: HsT18361, MGC2820, MRG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56075