Learn More
Selleck Chemical LLC MEISi-2 Dihydrochloride
SDP
Catalog No. 5000004287
Encompass_Preferred
MEISi-2 Dihydrochloride is a potent MEIS inhibitor (MEISi) that significantly inhibits MEIS-luciferase reporters in vitro The solubility of this product is not good please choose carefully for cell experiments
Catalog No. 50-000-04287 Supplier Selleck Chemical LLC Supplier No. S60625MG
Add to Cart
Add to Cart
MEISi-2 Dihydrochloride is a potent MEIS inhibitor (MEISi) that significantly inhibits MEIS-luciferase reporters in vitro The solubility of this product is not good please choose carefully for cell experiments