Learn More
Cayman Chemical MeIsoIndIgo 10mg
SDP
Catalog No. 502980103
Encompass_Preferred
An anticancer agent; inhibits the growth of NB4, HL-60, and U937 leukemia and primary acute myeloid leukemia cells at 5 and 10 µM; induces apoptosis in leukemia cell lines, primary AML cells, and CML cells; decreases spleen size in an HL-60 mouse xenograft model of AML in a dose-dependent manner at 50-150 mg/kg
Catalog No. 50-298-0103 Supplier Cayman Chemical Supplier No. 1800710 MG
An anticancer agent; inhibits the growth of NB4, HL-60, and U937 leukemia and primary acute myeloid leukemia cells at 5 and 10 µM; induces apoptosis in leukemia cell lines, primary AML cells, and CML cells; decreases spleen size in an HL-60 mouse xenograft model of AML in a dose-dependent manner at 50-150 mg/kg