Learn More
Medchemexpress LLC HY-13680 5mg Medchemexpress, Meisoindigo CAS:97207-47-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136805MG
Encompass_Preferred
Medchemexpress, HY-13680 5mg Meisoindigo CAS:97207-47-1 Meisoindigo (Dian III), a derivative of Indirubin (HY-N0117), halts the cell cycle at the G0/G1 phase and induces apoptosis in primary acute myeloid leukemia (AML) cells. Meisoindigo exhibits high antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1461