Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Babcock Intestinal Forceps

Catalog No. 12460592
Change view
Click to view available options
Length (Metric):
159 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
12-460-592 159 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-460-592 Supplier Integra™ Miltex™ Supplier No. MH1644
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Stainless steel
  • Non-glare mattte finish

Specifications

Product Type Forceps, Intestinal, Babcock
Material Stainless Steel
Finish Non-glare Matte
For Use With (Application) Intestinal surgery
Quantity Each
Tip Style Wide
Length (English) 6.25 in.
Length (Metric) 159 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.