Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Backhaus Towel Clamps

Catalog No. 12460716
Change view
Click to view available options
Length (Metric):
10.2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
12-460-716 10.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-460-716 Supplier Integra™ Miltex™ Supplier No. MH7504
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fixes drapes to the skin of anesthetized patients

  • Stainless steel
  • Length: 3.5 in. (8.9cm)
  • Non-glare matte finish

Specifications

Product Type Standard grade backhaus towel clamp
Length (English) 4 in.
Length (Metric) 10.2 cm
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.