Learn More
Integra™ Miltex™ MeisterHand™ Bishop-Harmon Micro Tissue Forceps
Catalog No. 12460616
Change view
Click to view available options
Length (Metric):
86 mm
Catalog No. Length (Metric)
12-460-616 86 mm
1 options
Catalog No. 12-460-616 Supplier Integra™ Miltex™ Supplier No. MH18867
Add to Cart
Add to Cart
Delicate tip design
  • Stainless steel
  • Fenestrated handle
  • Non-glare matte finish
Specifications
Product Type Forceps, Micro, Tissue, Bishop-Harmon
Material Stainless Steel
Finish Non-glare Matte
Quantity Each
Teeth Count 1 x 2
Length (English) 3.375 in.
Length (Metric) 86 mm