Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Halsted Mosquito Forceps

Catalog No. p-4185004
Change view
Click to view available options
Tip Style:
Curved
Curved, Extra Delicate
Straight
Straight, Fine
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
12-460-703 Curved, Extra Delicate
12-460-707 Straight
12-460-712 Curved
12-460-720 Straight, Fine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-460-703 Supplier Integra™ Miltex™ Supplier No. MH710
Only null left
Add to Cart
Add to Cart

Pattern length 4.75 in. (12.1cm)

  • Stainless steel
  • Non-glare matte finish
  • Full serrated jaws

Specifications

Tip Style Curved, Extra Delicate
Length (English) 4.75 in.
Material Stainless Steel
Length (Metric) 120 mm
Product Type Forceps, Mosquito, Halsted
Quantity (No. per Case) Each
Finish Non-glare Matte
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.