Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Heiss Self-Retaining Cross Action RetractorsRetractors

Catalog No. 12460588
Change view
Click to view available options
Length (English):
4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
12-460-588 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-460-588 Supplier Integra™ Miltex™ Supplier No. MH1116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blunt prongs

  • Surgical-grade German stainless steel with unique matte finish
  • Length: 4.125 in. (10.6cm)

Specifications

Product Type Cross-action Retractor
Grade Standard
Length (English) 4 in.
Length (Metric) 10.2 cm
Material Stainless Steel
Finish Matte
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.