Learn More
Integra™ Miltex™ MeisterHand™ Olsen-Hegar Needle Holders
Catalog No. 12460724
Change view
Click to view available options
Length (Metric):
14 cm
16.5 cm
19 cm
Material:
Stainless Steel
Tungsten Carbide
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Material
12-460-724 16.5 cm Tungsten Carbide
12-460-723 14 cm Tungsten Carbide
12-460-722 14 cm Stainless Steel
12-460-725 19 cm Tungsten Carbide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-460-724 Supplier Integra™ Miltex™ Supplier No. MH816TC
Add to Cart
Add to Cart
With suture scissors
  • Serrated jaws
  • Non-glare matte finish
  • Tungsten carbide jaw inserts can be replaced
  • 2500 teeth per square ince on tungsten carbide jaws
Specifications
Product Type Olsen-Hegar Needle Holder
Material Tungsten Carbide
Includes Suture Scissor
Style Serrated
Length (Metric) 16.5 cm