Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Stille-Liston Bone Cutting Forceps

Catalog No. 12460623
Catalog No. 12-460-623 Supplier Integra™ Miltex™ Supplier No. MH25390
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Straight design

  • Non-glare matte finish
  • Stainless-steel

Specifications

Product Type Forceps, Bone Cutting, Stille-Liston
Material Stainless Steel
Finish Matte
For Use With (Application) Bone cutting
Grade Surgical
Quantity Each
Tip Style Straight
Length (English) 10.75 in.
Length (Metric) 273 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.