Learn More
Selleck Chemical LLC Binimetinib (MEK162) 10mg 606143-89-9 ARRY-162,ARRY-438162
SDP
Catalog No. NC0660423
Encompass_Preferred
Binimetinib (MEK162, ARRY-162, ARRY-438162) is a potent inhibitor of MEK1/2 with IC50 of 12 nM in a cell-free assay. Binimetinib induces G1 cell cycle arrest and apoptosis in human NSCLC cell lines and induces autophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0660423 Supplier Selleck Chemical LLC Supplier No. S700710MG
Binimetinib (MEK162, ARRY-162, ARRY-438162) is a potent inhibitor of MEK1/2 with IC50 of 12 nM in a cell-free assay. Binimetinib induces G1 cell cycle arrest and apoptosis in human NSCLC cell lines and induces autophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.