Learn More
Enzo Life Sciences MEK1/2 (pSer218/222/226) monoclonal antibody (7E10) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905652100
Encompass_Preferred
Clone: 7E10. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of MEK1 phosphorylated at Ser218/222and MEK2 phosphorylated at Ser222/226 , conjugated to KLH. UniProt ID: Q02750 (MEK1), P36507 (MEK2). Species reactivity: Human, Mouse, Dog. Applications: ELISA, WB. Recommended Dilutions: ELISA (0.1µg/ml) Western Blot (0.5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-6985