Learn More
Cayman Chemical MEK1/2 (Phospho-Ser218,222) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502886657
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Ser218,222 of human MEK1/2 • Host: rabbit • Cross Reactivity: (+) NIH 3T3 cells • Application(s): WB • MEK1 is an integral component of the MAPK cascade that regulates cell growth and differentiation.
Catalog No. 50-288-6657 Supplier Cayman Chemical Supplier No. 100091781 EA
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Ser218,222 of human MEK1/2 • Host: rabbit • Cross Reactivity: (+) NIH 3T3 cells • Application(s): WB • MEK1 is an integral component of the MAPK cascade that regulates cell growth and differentiation.