Learn More
Eurofins Discoverx MEK1, active, 10 µg. (14-429)
Supplier:  Eurofins Discoverx 14429
Encompass
N-terminal GST and C-terminal 6xHis-tagged recombinant, human full length MEK1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8793