Learn More
Eurofins Discoverx MEK1, unactive, 50 µg. (14-420)
Supplier:  Eurofins Discoverx 14420
Encompass
N-terminal GST-tagged and C-terminal 6xHis-tagged recombinant, human full length MEK1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8791